Assay Details
| Gene Symbol: ARL14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031761.3 | NP_001026931.2 | 1 - 2 | 148 | 147 | |
| GenBank mRNA | BC112569.1 | - | 1 - 2 | 151 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112569.1, NM_001031761.3 transcripts.
Set Membership: |
|

ARL14EP
Genomic Map