Assay Details
| Gene Symbol: ARL14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031761.3 | NP_001026931.2 | 3 - 4 | 767 | 118 | |
| GenBank mRNA | BC112569.1 | - | 3 - 4 | 770 | 118 | |
| BT020677.1 | - | 3 - 4 | 700 | 118 | ||
More Information
Set Membership: |
|

ARL14EP
Genomic Map