Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034310.2 | NP_001029482.1 | 955 | 83 | ||
| GenBank mRNA | BC102656.1 | - | 3 - 4 | 394 | 83 | |
| BC111142.1 | - | 955 | 83 | |||
More Information
Set Membership: |
|

PTTG1
Genomic Map