Assay Details
| Gene Symbol: BUD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034457.2 | NP_001029629.2 | 1 - 2 | 76 | 107 | |
| GenBank mRNA | BC109889.1 | - | 1 - 2 | 76 | 107 | |
| BT021557.1 | - | 1 - 2 | 101 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109889.1, NM_001034457.2 transcripts.
Set Membership: |
|

BUD23
Genomic Map