Assay Details
| Gene Symbol: CHMP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191260.2 | NP_001178189.1 | 3 - 4 | 225 | 83 | |
| GenBank mRNA | BC109956.1 | - | 3 - 4 | 226 | 83 | |
| DT837819.1 | - | 3 - 4 | 226 | 83 | ||
More Information
Set Membership: |
|

CHMP1A
Genomic Map