Assay Details
| Gene Symbol: GID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037587.2 | NP_001032676.1 | 1 - 2 | 64 | 122 | |
| GenBank mRNA | BC109762.1 | - | 1 - 2 | 65 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109762.1, NM_001037587.2 transcripts.
Set Membership: |
|

GID8
Genomic Map