Assay Details
| Gene Symbol: GID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037587.2 | NP_001032676.1 | 3 - 4 | 392 | 70 | |
| GenBank mRNA | BC109762.1 | - | 3 - 4 | 393 | 70 | |
More Information
Set Membership: |
|

GID8
Genomic Map