Assay Details
| Gene Symbol: MARCHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037589.1 | NP_001032678.1 | 4 - 5 | 595 | 68 | |
| GenBank mRNA | BC109744.1 | - | 4 - 5 | 595 | 68 | |
More Information
Set Membership: |
|

MARCHF2
Genomic Map