Assay Details
| Gene Symbol: RMDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037600.2 | NP_001032689.1 | 2 - 3 | 307 | 119 | |
| GenBank mRNA | BC110037.1 | - | 2 - 3 | 308 | 119 | |
More Information
Set Membership: |
|

RMDN1
Genomic Map