Assay Details
| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037604.2 | NP_001032693.1 | 2 - 3 | 417 | 95 | |
| GenBank mRNA | BC109978.1 | - | 2 - 3 | 420 | 95 | |
More Information
Set Membership: |
|

GLRX
Genomic Map