Assay Details
| Gene Symbol: PSMB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037612.2 | NP_001032701.1 | 1 - 2 | 547 | 84 | |
| GenBank mRNA | BC110039.1 | - | 1 - 2 | 548 | 84 | |
More Information
Set Membership: |
|

PSMB5
Genomic Map