Assay Details
| Gene Symbol: HMGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037616.1 | NP_001032705.1 | 3 - 4 | 428 | 59 | |
| GenBank mRNA | BC109755.1 | - | 3 - 4 | 428 | 59 | |
More Information
Set Membership: |
|

HMGB2
Genomic Map