Assay Details
| Gene Symbol: C10H14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038049.1 | NP_001033138.1 | 5 - 6 | 1433 | 74 | |
| GenBank mRNA | BC111308.1 | - | 5 - 6 | 1433 | 74 | |
More Information
Set Membership: |
|

C10H14orf93
Genomic Map