Assay Details
| Gene Symbol: IFI27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038050.2 | NP_001033139.1 | 2 - 3 | 235 | 88 | |
| GenBank mRNA | BC111178.1 | - | 2 - 3 | 236 | 88 | |
More Information
Set Membership: |
|

IFI27
Genomic Map