Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038055.2 | NP_001033144.1 | 5 - 6 | 705 | 79 | |
| GenBank mRNA | BC111170.1 | - | 5 - 6 | 706 | 79 | |
More Information
Set Membership: |
|

KNSTRN
Genomic Map