Assay Details
| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038056.2 | NP_001033145.1 | 2 - 3 | 535 | 112 | |
| GenBank mRNA | BC111135.1 | - | 2 - 3 | 536 | 112 | |
More Information
Set Membership: |
|

TM2D2
Genomic Map