Assay Details
| Gene Symbol: FBXO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038059.1 | NP_001033148.1 | 8 - 9 | 1459 | 67 | |
| GenBank mRNA | BC111286.1 | - | 8 - 9 | 1459 | 67 | |
More Information
Set Membership: |
|

FBXO7
Genomic Map