Assay Details
| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038065.1 | NP_001033154.1 | 1 - 2 | 309 | 60 | |
| GenBank mRNA | BC111183.1 | - | 1 - 2 | 309 | 60 | |
More Information
Set Membership: |
|

GPNMB
Genomic Map