Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038067.1 | NP_001033156.1 | 5 - 6 | 2073 | 70 | |
| GenBank mRNA | BC109748.1 | - | 5 - 6 | 2073 | 70 | |
More Information
Set Membership: |
|

CABYR
Genomic Map