Assay Details
| Gene Symbol: MAP2K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038071.2 | NP_001033160.2 | 10 - 11 | 1355 | 57 | |
| GenBank mRNA | BC118362.1 | - | 10 - 11 | 1357 | 57 | |
More Information
Set Membership: |
|

MAP2K2
Genomic Map