Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038074.1 | NP_001033163.1 | 4 - 5 | 697 | 55 | |
| GenBank mRNA | BC110035.1 | - | 4 - 5 | 697 | 55 | |
More Information
Set Membership: |
|

EMC8
Genomic Map