Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038080.2 | NP_001033169.1 | 5 - 6 | 728 | 66 | |
| GenBank mRNA | BC109953.1 | - | 5 - 6 | 729 | 66 | |
More Information
Set Membership: |
|

STK17B
Genomic Map