Assay Details
| Gene Symbol: NINJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038082.2 | NP_001033171.1 | 2 - 3 | 377 | 69 | |
| GenBank mRNA | BC111141.1 | - | 2 - 3 | 378 | 69 | |
More Information
Set Membership: |
|

NINJ1
Genomic Map