Assay Details
| Gene Symbol: MED11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038084.2 | NP_001033173.1 | 2 - 3 | 280 | 127 | |
| GenBank mRNA | BC109828.1 | - | 2 - 3 | 281 | 127 | |
| BC149739.1 | - | 1 - 2 | 415 | 127 | ||
More Information
Set Membership: |
|

MED11
Genomic Map