Assay Details
| Gene Symbol: CYC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038090.2 | NP_001033179.1 | 1 - 2 | 169 | 55 | |
| GenBank mRNA | BC109917.1 | - | 1 - 2 | 170 | 55 | |
More Information
Set Membership: |
|

CYC1
Genomic Map