Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038091.2 | NP_001033180.1 | 12 - 13 | 1590 | 78 | |
| GenBank mRNA | BC111362.1 | - | 12 - 13 | 1591 | 78 | |
More Information
Set Membership: |
|

MARS1
Genomic Map