Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038091.2 | NP_001033180.1 | 17 - 18 | 2259 | 76 | |
| GenBank mRNA | BC111362.1 | - | 17 - 18 | 2260 | 76 | |
More Information
Set Membership: |
|

MARS1
Genomic Map