Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038103.2 | NP_001033192.1 | 5 - 6 | 780 | 54 | |
| GenBank mRNA | BC111312.1 | - | 5 - 6 | 781 | 54 | |
More Information
Set Membership: |
|

TEKT2
Genomic Map