Assay Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038105.2 | NP_001033194.1 | 3 - 4 | 268 | 64 | |
| GenBank mRNA | BC111309.1 | - | 3 - 4 | 269 | 64 | |
More Information
Set Membership: |
|

RSPH1
Genomic Map