Assay Details
| Gene Symbol: POLR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038124.2 | NP_001033213.1 | 2 - 3 | 199 | 110 | |
| GenBank mRNA | BC109803.1 | - | 2 - 3 | 200 | 110 | |
More Information
Set Membership: |
|

POLR1C
Genomic Map