Assay Details
| Gene Symbol: GCLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038143.1 | NP_001033232.1 | 2 - 3 | 434 | 69 | |
| GenBank mRNA | BC111206.1 | - | 2 - 3 | 434 | 69 | |
More Information
Set Membership: |
|

GCLM
Genomic Map