Assay Details
| Gene Symbol: TUBA3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038163.2 | NP_001033252.1 | 2 - 3 | 298 | 76 | |
| GenBank mRNA | BC110002.1 | - | 2 - 3 | 301 | 76 | |
More Information
Set Membership: |
|

TUBA3E
Genomic Map