Assay Details
| Gene Symbol: PGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038172.2 | NP_001033261.1 | 1 - 2 | 717 | 77 | |
| GenBank mRNA | BC111291.1 | - | 1 - 2 | 697 | 77 | |
More Information
Set Membership: |
|

PGP
Genomic Map