Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038174.2 | NP_001033263.1 | 2 - 3 | 402 | 84 | |
| GenBank mRNA | BC109497.1 | - | 2 - 3 | 403 | 84 | |
More Information
Set Membership: |
|

RNF138
Genomic Map