Assay Details
| Gene Symbol: C23H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038178.2 | NP_001033267.1 | 1 - 2 | 172 | 84 | |
| GenBank mRNA | BC109937.1 | - | 1 - 2 | 173 | 84 | |
More Information
Set Membership: |
|

C23H6orf136
Genomic Map