Assay Details
| Gene Symbol: C23H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038178.2 | NP_001033267.1 | 4 - 5 | 858 | 97 | |
| GenBank mRNA | BC109937.1 | - | 4 - 5 | 859 | 97 | |
More Information
Set Membership: |
|

C23H6orf136
Genomic Map