Assay Details
| Gene Symbol: SPATA22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038186.2 | NP_001033275.1 | 3 - 4 | 325 | 108 | |
| GenBank mRNA | BC111158.1 | - | 3 - 4 | 326 | 108 | |
More Information
Set Membership: |
|

SPATA22
Genomic Map