Assay Details
| Gene Symbol: JHY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038208.1 | NP_001033297.1 | 1 - 2 | 117 | 82 | |
| GenBank mRNA | BC111348.1 | - | 1 - 2 | 117 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111348.1, NM_001038208.1 transcripts.
Set Membership: |
|

JHY
Genomic Map