Assay Details
| Gene Symbol: PMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038212.2 | NP_001033301.1 | 4 - 5 | 595 | 137 | |
| GenBank mRNA | BC111342.1 | - | 4 - 5 | 596 | 137 | |
More Information
Set Membership: |
|

PMF1
Genomic Map