Assay Details
| Gene Symbol: CORO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038220.1 | NP_001033309.1 | 11 - 12 | 1536 | 90 | |
| GenBank mRNA | BC109477.1 | - | 11 - 12 | 1536 | 90 | |
More Information
Set Membership: |
|

CORO2A
Genomic Map