Assay Details
| Gene Symbol: CALR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038514.2 | NP_001033603.1 | 8 - 9 | 1056 | 92 | |
| GenBank mRNA | BC109750.1 | - | 8 - 9 | 1057 | 92 | |
More Information
Set Membership: |
|

CALR3
Genomic Map