Assay Details
| Gene Symbol: VBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038516.2 | NP_001033605.1 | 4 - 5 | 428 | 78 | |
| GenBank mRNA | BC109526.1 | - | 4 - 5 | 429 | 78 | |
More Information
Set Membership: |
|

VBP1
Genomic Map