Assay Details
| Gene Symbol: SNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038519.1 | NP_001033608.1 | 6 - 7 | 691 | 68 | |
| GenBank mRNA | BC109562.1 | - | 6 - 7 | 691 | 68 | |
More Information
Set Membership: |
|

SNX2
Genomic Map