Assay Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038539.2 | NP_001033628.1 | 3 - 4 | 343 | 83 | |
| GenBank mRNA | BC109494.1 | - | 3 - 4 | 344 | 83 | |
More Information
Set Membership: |
|

PSMB1
Genomic Map