Assay Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038546.2 | NP_001033635.1 | 2 - 3 | 208 | 67 | |
| GenBank mRNA | BC109999.1 | - | 2 - 3 | 210 | 67 | |
More Information
Set Membership: |
|

SNUPN
Genomic Map