Assay Details
| Gene Symbol: PAQR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038553.1 | NP_001033642.1 | 1 - 2 | 99 | 78 | |
| GenBank mRNA | BC111284.1 | - | 1 - 2 | 99 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111284.1, NM_001038553.1 transcripts.
Set Membership: |
|

PAQR7
Genomic Map