Assay Details
| Gene Symbol: NT5C1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038555.1 | NP_001033644.1 | 8 - 9 | 1416 | 76 | |
| GenBank mRNA | BC109780.1 | - | 8 - 9 | 1416 | 76 | |
More Information
Set Membership: |
|

NT5C1B
Genomic Map