Assay Details
| Gene Symbol: MMP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038556.2 | NP_001033645.1 | 4 - 5 | 618 | 110 | |
| GenBank mRNA | BC109902.1 | - | 4 - 5 | 620 | 110 | |
More Information
Set Membership: |
|

MMP23
Genomic Map