Assay Details
| Gene Symbol: POC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038560.2 | NP_001033649.1 | 10 - 11 | 1168 | 59 | |
| GenBank mRNA | BC109862.1 | - | 10 - 11 | 1169 | 59 | |
More Information
Set Membership: |
|

POC1A
Genomic Map