Assay Details
| Gene Symbol: ARL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038567.2 | NP_001033656.1 | 4 - 5 | 343 | 88 | |
| GenBank mRNA | BC109565.1 | - | 4 - 5 | 344 | 88 | |
More Information
Set Membership: |
|

ARL3
Genomic Map